3-Chloro-α,α-difluorobenzeneacetic acid 95%

Supplier: Ambeed
Warning

A155672-100MG A155672-250MG A155672-5G A155672-1G
77150-636EA 16 USD
77150-636 77150-638 77150-640 77901-102
3-Chloro-α,α-difluorobenzeneacetic acid 95%
3-Chloro-α,α-difluorobenzeneacetic acid

Precaución: for Research Use Only

Formula: C₈H₅ClF₂O₂
MDL Number: MFCD11007696
CAS Number: 1027513-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR