cis-tert-Butyl 3-hydroxy-3-methylcyclobutylcarbamate 98%

Supplier: Ambeed
Warning

A155037-1G A155037-250MG A155037-5G
77150-628EA 126.69 USD
77150-628 77150-630 77150-632
cis-tert-Butyl 3-hydroxy-3-methylcyclobutylcarbamate 98%
cis-tert-Butyl 3-hydroxy-3-methylcyclobutylcarbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₃
MW: 201.26 g/mol
MDL Number: MFCD20926191
CAS Number: 1363381-12-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR