Creatine phosphate disodium salt tetrahydrate 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
Sodium creatine phosphate dibasic tetrahydrate, Sodium N-carbamimidoyl-N-methylglycinate dihydrogen phosphate tetrahydrate, Phosphocreatine disodium salt tetrahydrate
A154854-25G
A154854-5G
A154854-100G
77150-616EA
60.77
USD
77150-616
77150-618
77685-278
Creatine phosphate disodium salt tetrahydrate 99%
Creatine phosphate disodium salt tetrahydrate
Precaución: for Research Use Only
Formula:
C₄H₈N₃Na₂O₅P·4H₂O MW: 327.14 g/mol (255.07 anhy) |
MDL Number:
MFCD00150192 CAS Number: 71519-72-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...