Fmoc-Ser(BSi)-OH 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Ser(TBDMS)-OH

A154547-10G A154547-1G A154547-250MG A154547-25G A154547-5G
77150-590EA 154.5 USD
77150-590 77150-592 77150-594 77150-596 77150-598
Fmoc-Ser(BSi)-OH 97%
Fmoc-Ser(BSi)-OH

Precaución: for Research Use Only

Formula: C₂₄H₃₁NO₅Si
MW: 441.599 g/mol
MDL Number: MFCD02092931
CAS Number: 146346-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR