2-(4-Bromophenyl)benzothiazole 98%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Benzothiazolyl)-4-bromobenzene

A154494-100G A154494-10G A154494-25G A154494-5G
77150-582EA 241.02 USD
77150-582 77150-584 77150-586 77150-588
2-(4-Bromophenyl)benzothiazole 98%
2-(4-Bromophenyl)benzothiazole

Precaución: for Research Use Only

Formula: C₁₃H₈BrNS
MW: 290.18 g/mol
MDL Number: MFCD00579199
CAS Number: 19654-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR