3,5-Dichloroisonicotinic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dichloropyridine-4-carboxylic acid

A154264-10G A154264-1G A154264-250MG A154264-5G
77150-564EA 47.38 USD
77150-564 77150-566 77150-568 77150-570
3,5-Dichloroisonicotinic acid 97%
3,5-Dichloroisonicotinic acid

Precaución: for Research Use Only

Formula: C₆H₃Cl₂NO₂
MW: 192 g/mol
MDL Number: MFCD01861975
CAS Number: 13958-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR