2,4-diaminopyrimidin-5-ol dihydrochloride 95%

Supplier: Ambeed
Warning

A154061-100MG A154061-1G A154061-250MG
77150-544EA 42.23 USD
77150-544 77150-546 77150-548
2,4-diaminopyrimidin-5-ol dihydrochloride 95%
2,4-diaminopyrimidin-5-ol dihydrochloride

Precaución: for Research Use Only

Formula: C₄H₈Cl₂N₄O
MW: 199.04 g/mol
MDL Number: MFCD29044857
CAS Number: 141124-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR