4-(Methylsulfonyl)-2-(trifluoromethyl)benzaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 4-Formyl-3-(trifluoromethyl)phenyl methyl sulphone, 2-Formyl-5-(methylsulphonyl)benzotrifluoride

A153870-1G A153870-250MG A153870-25G A153870-5G
77150-522EA 19.57 USD
77150-522 77150-524 77150-526 77150-528
4-(Methylsulfonyl)-2-(trifluoromethyl)benzaldehyde 97%
4-(Methylsulfonyl)-2-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₇F₃O₃S
MW: 252.21 g/mol
MDL Number: MFCD15479075
CAS Number: 1215310-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR