5-Bromo-1,2,3,4-tetrahydroquinoline hydrochloride 97%

Supplier: Ambeed
Warning

A153868-100MG A153868-1G A153868-250MG A153868-5G
77150-514EA 7 USD
77150-514 77150-516 77150-518 77150-520
5-Bromo-1,2,3,4-tetrahydroquinoline hydrochloride 97%
5-Bromo-1,2,3,4-tetrahydroquinoline hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₁BrClN
MDL Number: MFCD09026771
CAS Number: 1073968-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR