2-((5-Chloropyridin-2-yl)amino)-2-oxoacetic acid 97%

Supplier: Ambeed
Danger

A153705-100G A153705-10G A153705-250MG A153705-25G A153705-5G
77150-488EA 288.4 USD
77150-488 77150-490 77150-492 77150-494 77150-496
2-((5-Chloropyridin-2-yl)amino)-2-oxoacetic acid 97%
2-((5-Chloropyridin-2-yl)amino)-2-oxoacetic acid

Precaución: for Research Use Only

Formula: C₇H₅ClN₂O₃
MW: 200.58 g/mol
MDL Number: MFCD12171750
CAS Number: 552850-73-4
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR