trans-4-Aminocyclohexanol hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A153433-100G
A153433-10G
A153433-25G
A153433-5G
A153433-500G
77150-474EA
44
USD
77150-474
77150-476
77150-478
77150-480
77567-226
trans-4-Aminocyclohexanol hydrochloride 97%
trans-4-Aminocyclohexanol hydrochloride
Precaución: For research use only.
Formula:
C₆H₁₄ClNO |
MDL Number:
MFCD00012566 CAS Number: 50910-54-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...