N-Acetyl-L-leucine 95%

Supplier: Ambeed
Warning

Synonyms: N-Ac-Leu-OH, Ac-Leu-OH

A153237-1000G A153237-100G A153237-10G A153237-25G A153237-500G
77150-466EA 75 USD
77150-466 77150-468 77150-470 77150-472 77685-267
N-Acetyl-L-leucine 95%
N-Acetyl-L-leucine

Precaución: for Research Use Only

Formula: C₈H₁₅NO₃
MW: 173.2 g/mol
MDL Number: MFCD00065131
CAS Number: 1188-21-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR