CEP-33779 98%

Supplier: Ambeed
Warning

Synonyms: N-(3-(4-Methylpiperazin-1-yl)phenyl)-8-(4-(methylsulphonyl)phenyl)-[1,2,4]triazolo[1,5-a]pyridin-2-amine

A153169-10MG A153169-1MG A153169-50MG A153169-5MG
77150-458EA 50.47 USD
77150-458 77150-460 77150-462 77150-464
CEP-33779 98%
CEP-33779

Precaución: for Research Use Only

Formula: C₂₄H₂₆N₆O₂S
MW: 462.57 g/mol
MDL Number: MFCD22683932
CAS Number: 1257704-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR