ent-Cinacalcet Hydrochloride 97%

Supplier: Ambeed
Warning

A152647-100MG A152647-1G A152647-250MG
77150-452EA 83.43 USD
77150-452 77150-454 77150-456
ent-Cinacalcet Hydrochloride 97%
ent-Cinacalcet Hydrochloride

Precaución: for Research Use Only

Formula: C₂₂H₂₃ClF₃N
MW: 393.87 g/mol
MDL Number: MFCD23160290
CAS Number: 1217809-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR