Tenovin-1 98%

Supplier: Ambeed
Warning

A152615-100MG A152615-10MG A152615-1MG A152615-50MG A152615-5MG
77150-442EA 180 USD
77150-442 77150-444 77150-446 77150-448 77150-450
Tenovin-1 98%
Tenovin-1

Precaución: for Research Use Only

Formula: C₂₀H₂₃N₃O₂S
MDL Number: MFCD02914760
CAS Number: 380315-80-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR