4-Bromo-2-chloro-1-iodobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-3-chloro-4-iodobenzene

A152478-100G A152478-10G A152478-500G A152478-5G A152478-25G
77150-434EA 71.07 USD
77150-434 77150-436 77150-438 77150-440 77318-184
4-Bromo-2-chloro-1-iodobenzene 98%
4-Bromo-2-chloro-1-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrClI
MW: 317.35 g/mol
MDL Number: MFCD00079709
CAS Number: 31928-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR