(S)-2-Amino-3-(3-fluorophenyl)-2-methylpropanoic acid 95%

Supplier: Ambeed
Warning

A152453-100MG A152453-1G A152453-250MG
77150-428EA 52.53 USD
77150-428 77150-430 77150-432
(S)-2-Amino-3-(3-fluorophenyl)-2-methylpropanoic acid 95%
(S)-2-Amino-3-(3-fluorophenyl)-2-methylpropanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂FNO₂
MW: 197.21 g/mol
MDL Number: MFCD18695499
CAS Number: 130855-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR