4-Chloro-2-methyl-1-(phenylsulfonyl)-1H-pyrrolo[2,3-b]pyridine 95%

Supplier: Ambeed
Warning

A152048-100MG A152048-1G A152048-250MG A152048-50MG A152048-5G
77150-408EA 150.38 USD
77150-408 77150-410 77150-412 77150-414 77150-416
4-Chloro-2-methyl-1-(phenylsulfonyl)-1H-pyrrolo[2,3-b]pyridine 95%
4-Chloro-2-methyl-1-(phenylsulfonyl)-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₁₄H₁₁ClN₂O₂S
MW: 306.76 g/mol
MDL Number: MFCD15529557
CAS Number: 744209-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR