Fmoc-Tyr-OtBu 95%

Supplier: Ambeed
Warning

A151978-100G A151978-10G A151978-1G A151978-25G A151978-500G A151978-5G
77150-396EA 393.46 USD
77150-396 77150-398 77150-400 77150-402 77150-404 77150-406
Fmoc-Tyr-OtBu 95%
Fmoc-Tyr-OtBu

Precaución: for Research Use Only

Formula: C₂₈H₂₉NO₅
MW: 459.5 g/mol
MDL Number: MFCD20264831
CAS Number: 133852-23-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR