Ethyl-trans-4-decenoate 98%

Supplier: Ambeed
Warning

Synonyms: trans-Obtusilic Acid Ethyl Ester, Ethyl trans-4-Decenoate, trans-4-Decenoic Acid Ethyl Ester

A151682-5G A151682-1G
77150-386EA 20.6 USD
77150-386 77611-414
Ethyl-trans-4-decenoate 98%
Ethyl-trans-4-decenoate

Precaución: for Research Use Only

Formula: C₁₂H₂₂O₂
MW: 198.31 g/mol
MDL Number: MFCD00015574
CAS Number: 76649-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR