2-((2S,6R)-6-(((S)-1-Ethoxy-1-oxo-4-phenylbutan-2-yl)amino)-5-oxo-2-(thiophen-2-yl)-1,4-thiazepan-4-yl)acetic acid hydrochloride 98%
Supplier: Ambeed
|
Warning
|
A151541-10MG
A151541-1MG
A151541-50MG
77150-380EA
44
USD
77150-380
77150-382
77150-384
2-((2S,6R)-6-(((S)-1-Ethoxy-1-oxo-4-phenylbutan-2-yl)amino)-5-oxo-2-(thiophen-2-yl)-1,4-thiazepan-4-yl)acetic acid hydrochloride 98%
2-((2S,6R)-6-(((S)-1-Ethoxy-1-oxo-4-phenylbutan-2-yl)amino)-5-oxo-2-(thiophen-2-yl)-1,4-thiazepan-4-yl)acetic acid hydrochloride
Precaución: for Research Use Only
Formula:
C₂₃H₂₉ClN₂O₅S₂ |
MDL Number:
MFCD00933433 CAS Number: 110221-44-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...