K02288 98%

Supplier: Ambeed
Danger

Synonyms: 3-[6-Amino-5-(3,4,5-trimethoxyphenyl)pyridin-3-yl]phenol

A151373-10MG A151373-1MG A151373-50MG
77150-368EA 66.95 USD
77150-368 77150-370 77150-372
K02288 98%
K02288

Precaución: for Research Use Only

Formula: C₂₀H₂₀N₂O₄
MW: 352.38 g/mol
MDL Number: MFCD26936347
CAS Number: 1431985-92-0
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR