2-Bromo-5-(methylthio)pyridine 96%

Supplier: Ambeed
Warning

A150224-100MG A150224-250MG A150224-1G
77150-354EA 15 USD
77150-354 77150-356 77901-440
2-Bromo-5-(methylthio)pyridine 96%
2-Bromo-5-(methylthio)pyridine

Precaución: for Research Use Only

Formula: C₆H₆BrNS
MDL Number: MFCD04974522
CAS Number: 134872-23-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR