Pentafluoroanilinium trifluoromethanesulfonate 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
2,3,4,5,6-Pentafluoroaniline trifluoromethanesulphonate, Pentafluoroanilinium Triflate, PFPAT, (Pentafluorophenyl)ammonium Trifluoromethanesulphonate
A147785-5G
A147785-1G
77150-302EA
261
USD
77150-302
77900-847
Pentafluoroanilinium trifluoromethanesulfonate 97%
Pentafluoroanilinium trifluoromethanesulfonate
Precaución: for Research Use Only
Formula:
C₇H₃F₈NO₃S MW: 333.15 g/mol |
MDL Number:
MFCD08276416 CAS Number: 912823-79-1 UN: 1759 ADR: 8 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...