Ethyl-5-chloro-2-pyridinecarboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5-chloropicolinate

A147606-100MG A147606-1G A147606-250MG A147606-5G
77150-292EA 18.54 USD
77150-292 77150-294 77150-296 77150-298
Ethyl-5-chloro-2-pyridinecarboxylate 97%
Ethyl-5-chloro-2-pyridinecarboxylate

Precaución: for Research Use Only

Formula: C₈H₈ClNO₂
MW: 185.61 g/mol
MDL Number: MFCD07772018
CAS Number: 128072-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR