D(-)-allo-Isoleucine 97%

Supplier: Ambeed
Warning

Synonyms: H-D-Allo-Ile-OH, (2R,3S)-(-)-allo-Isoleucine, H-D-allo-Ile-OH

A146336-100G A146336-10G A146336-1G A146336-250MG A146336-25G A146336-5G
77150-262EA 519 USD
77150-262 77150-264 77150-266 77150-268 77150-270 77150-272
D(-)-allo-Isoleucine 97%
D(-)-allo-Isoleucine

Precaución: for Research Use Only

Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
MDL Number: MFCD00066445
CAS Number: 1509-35-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR