(R)-4-(2-Fluorophenyl)-3,4-dihydronaphthalen-1(2H)-one 98%

Supplier: Ambeed
Warning

A146256-100MG A146256-1G A146256-250MG
77150-256EA 40 USD
77150-256 77150-258 77150-260
(R)-4-(2-Fluorophenyl)-3,4-dihydronaphthalen-1(2H)-one 98%
(R)-4-(2-Fluorophenyl)-3,4-dihydronaphthalen-1(2H)-one

Precaución: for Research Use Only

Formula: C₁₆H₁₃FO
MDL Number: MFCD28963660
CAS Number: 1234356-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR