AG-120 (Ivosidenib) 99%

Supplier: Ambeed
Warning

Synonyms: IDH1 Inhibitor 8, (2S)-N-(1-(2-chlorophenyl)-2-((3,3-difluorocyclobutyl)amino)-2-oxoethyl)-1-(4-cyanopyridin-2-yl)-N-(5-fluoropyridin-3-yl)-5-oxopyrrolidine-2-carboxamide, Ivosidenib racemate

A145388-10MG A145388-1MG A145388-50MG A145388-5MG
77150-226EA 55.62 USD
77150-226 77150-228 77150-230 77150-232
AG-120 (Ivosidenib) 99%
AG-120 (Ivosidenib)

Precaución: for Research Use Only

Formula: C₂₈H₂₂ClF₃N₆O₆
MW: 582.97 g/mol
MDL Number: MFCD29044889
CAS Number: 1448347-49-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR