Methyl 2-chloro-4-methoxybenzoate 98%

Supplier: Ambeed
Warning

A145157-1G A145157-5G
77150-222EA 9 USD
77150-222 77150-224
Methyl 2-chloro-4-methoxybenzoate 98%
Methyl 2-chloro-4-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₉ClO₃
MDL Number: MFCD06797822
CAS Number: 104253-45-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR