3,5-Dibromoethylbenzene 95%

Supplier: Ambeed
Warning

A145099-1G A145099-250MG A145099-5G
77150-216EA 24 USD
77150-216 77150-218 77150-220
3,5-Dibromoethylbenzene 95%
3,5-Dibromoethylbenzene

Precaución: for Research Use Only

Formula: C₈H₈Br₂
MDL Number: MFCD11846036
CAS Number: 59785-43-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR