1-(2-Fluorophenyl)cyclopropanamine 96%

Supplier: Ambeed
Warning

A145061-100MG A145061-250MG A145061-5G A145061-1G
77150-210EA 13 USD
77150-210 77150-212 77150-214 77592-716
1-(2-Fluorophenyl)cyclopropanamine 96%
1-(2-Fluorophenyl)cyclopropanamine

Precaución: for Research Use Only

Formula: C₉H₁₀FN
MDL Number: MFCD07374467
CAS Number: 886366-50-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR