CEP-32496 ≥98%

Supplier: Ambeed
Warning

Synonyms: 1-(3-((6,7-Dimethoxyquinazolin-4-yl)oxy)phenyl)-3-(5-(1,1,1-trifluoro-2-methylpropan-2-yl)isoxazol-3-yl)urea, Agerafenib

A144993-10MG A144993-50MG A144993-5MG A144993-1MG
77150-204EA 6.18 USD
77150-204 77150-206 77150-208 77685-231
CEP-32496 ≥98%
CEP-32496

Precaución: for Research Use Only

Formula: C₂₄H₂₂F₃N₅O₅
MW: 517.47 g/mol
MDL Number: MFCD22124524
CAS Number: 1188910-76-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR