(S)-tert-Butyl (2,6-dimethyl-3-oxohept-1-en-4-yl)carbamate 96%

Supplier: Ambeed
Warning

A144520-1G A144520-250MG A144520-5G
77150-188EA 42.23 USD
77150-188 77150-190 77150-192
(S)-tert-Butyl (2,6-dimethyl-3-oxohept-1-en-4-yl)carbamate 96%
(S)-tert-Butyl (2,6-dimethyl-3-oxohept-1-en-4-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₄H₂₅NO₃
MW: 255.35 g/mol
MDL Number: MFCD26227261
CAS Number: 247068-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR