1-Methyl-2-(methylsulfanyl)-1H-imidazole 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Methyl-2-(methylthio)imidazole
A143808-10G
A143808-1G
A143808-250MG
A143808-5G
77150-180EA
437.75
USD
77150-180
77150-182
77150-184
77150-186
1-Methyl-2-(methylsulfanyl)-1H-imidazole 97%
1-Methyl-2-(methylsulfanyl)-1H-imidazole
Precaución: for Research Use Only
Formula:
C₅H₈N₂S MW: 128.195 g/mol |
MDL Number:
MFCD25959069 CAS Number: 14486-52-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...