3-Bromo-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazole 95%

Supplier: Ambeed
Warning

A143693-100MG A143693-250MG A143693-1G A143693-5G
77150-172EA 24 USD
77150-172 77150-174 77611-318 77685-217
3-Bromo-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazole 95%
3-Bromo-1-(tetrahydro-2H-pyran-2-yl)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₈H₁₁BrN₂O
MW: 231.08 g/mol
MDL Number: MFCD16659986
CAS Number: 1044270-96-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR