2-(Methoxycarbonyl)ethylboronic acid, pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)propanoate

A143309-1G A143309-250MG A143309-5G A143309-25G
77150-150EA 116.39 USD
77150-150 77150-152 77150-154 77150-156 77685-214
2-(Methoxycarbonyl)ethylboronic acid, pinacol ester 98%
2-(Methoxycarbonyl)ethylboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₀H₁₉BO₄
MDL Number: MFCD10700159
CAS Number: 1150561-77-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR