2-Methylindole 97%

Supplier: Ambeed
Warning

Synonyms: 2-Methyl-1H-indole

A143223-100G A143223-25G A143223-500G
77150-144EA 34 USD
77150-144 77150-146 77150-148
2-Methylindole 97%
2-Methylindole

Precaución: for Research Use Only

Formula: C₉H₉N
MDL Number: MFCD00005616
CAS Number: 95-20-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR