GSK2578215A 98%

Supplier: Ambeed
Warning

Synonyms: 2-(benzyloxy)-5-(2-fluoropyridin-4-yl)-N-(pyridin-3-yl)benzamide, LRRK2-kinase inhibitor, 5-(2-Fluoro-4-pyridinyl)-2-(phenylmethoxy)-N-3-pyridinylbenzamide

A142087-10MG A142087-1MG A142087-25MG A142087-5MG
77150-124EA 45.32 USD
77150-124 77150-126 77150-128 77611-296
GSK2578215A 98%
GSK2578215A

Precaución: for Research Use Only

Formula: C₂₄H₁₈FN₃O₂
MW: 399.42 g/mol
MDL Number: MFCD22665702
CAS Number: 1285515-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR