GSK-0660 98%

Supplier: Ambeed
Warning

Synonyms: methyl 3-[(4-anilino-2-methoxyphenyl)sulphamoyl]thiophene-2-carboxylate, GSK0660, 3-[[[2-Methoxy-4-(phenylamino)phenyl]amino]sulphonyl]-2-thiophenecarboxylic acid methyl ester, 3-[[[2-Methoxy-4-(phenylamino)phenyl]amino]sulphonyl]-2-thiophenecarboxylicacidmethylester, Methyl 3-(N-(2-methoxy-4-(phenylamino)phenyl)sulphamoyl)thiophene-2-carboxylate

A141734-10MG A141734-1MG A141734-50MG A141734-5MG
77150-096EA 100 USD
77150-096 77150-098 77150-100 77150-102
GSK-0660 98%
GSK-0660

Precaución: for Research Use Only

Formula: C₁₉H₁₈N₂O₅S₂
MDL Number: MFCD12828770
CAS Number: 1014691-61-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR