4'-Chloro-3-phenylpropiophenone 95%

Supplier: Ambeed
Warning

A141659-100MG A141659-1G A141659-250MG A141659-5G
77150-088EA 26.78 USD
77150-088 77150-090 77150-092 77150-094
4'-Chloro-3-phenylpropiophenone 95%
4'-Chloro-3-phenylpropiophenone

Precaución: for Research Use Only

Formula: C₁₅H₁₃ClO
MW: 244.72 g/mol
MDL Number: MFCD03842928
CAS Number: 5739-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR