Fmoc-Sar-OH 98% (by HPLC)
Supplier: Ambeed
|
Warning
|
Synonyms:
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methylglycine, N-Fmoc-N-methylglycine, Fmoc-N-Me-Gly-OH
A141362-100G
A141362-10G
A141362-25G
A141362-500G
A141362-5G
77150-074EA
41
USD
77150-074
77150-076
77150-078
77150-080
77150-082
Fmoc-Sar-OH 98% (by HPLC)
Fmoc-Sar-OH
Precaución: for Research Use Only
Formula:
C₁₈H₁₇NO₄ |
MDL Number:
MFCD00151926 CAS Number: 77128-70-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...