Boc-Dap(Fmoc)-OH 95%

Supplier: Ambeed
Warning

A141078-100G A141078-10G A141078-1G A141078-250MG A141078-25G A141078-5G
77150-062EA 196 USD
77150-062 77150-064 77150-066 77150-068 77150-070 77150-072
Boc-Dap(Fmoc)-OH 95%
Boc-Dap(Fmoc)-OH

Precaución: for Research Use Only

Formula: C₂₃H₂₆N₂O₆
MDL Number: MFCD00235897
CAS Number: 122235-70-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR