CGK733 98%

Supplier: Ambeed
Warning

A140835-10MG A140835-1MG A140835-50MG A140835-5MG
77150-054EA 47.99 USD
77150-054 77150-056 77150-058 77150-060
CGK733 98%
CGK733

Precaución: for Research Use Only

Formula: C₂₃H₁₈Cl₃FN₄O₃S
MDL Number: MFCD09038682
CAS Number: 905973-89-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR