Tideglusib 98%

Supplier: Ambeed
Warning

A140220-100MG A140220-10MG A140220-1MG A140220-50MG A140220-1G A140220-250MG A140220-5G
77150-034EA 41 USD
77150-034 77150-036 77150-038 77150-040 77566-818 77685-203 77685-204
Tideglusib 98%
Tideglusib

Precaución: For research use only.

Formula: C₁₉H₁₄N₂O₂S
MDL Number: MFCD18633296
CAS Number: 865854-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR