KPT-330 98%

Supplier: Ambeed
Warning

A140158-1MG A140158-5MG A140158-50MG A140158-100MG A140158-1G A140158-250MG
77150-030EA 22 USD
77150-030 77150-032 77685-202 77685-199 77685-200 77685-201
KPT-330 98%
KPT-330

Precaución: for Research Use Only

Formula: C₁₇H₁₁F₆N₇O
MDL Number: MFCD27987944
CAS Number: 1393477-72-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR