(2S,5R)-5-(4-((2-Fluorobenzyl)oxy)phenyl)pyrrolidine-2-carboxamide hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: Raxatrigine HCl

A139957-10MG A139957-1MG A139957-2MG A139957-5MG A139957-50MG
77150-018EA 138.02 USD
77150-018 77150-020 77150-022 77150-024 77611-276
(2S,5R)-5-(4-((2-Fluorobenzyl)oxy)phenyl)pyrrolidine-2-carboxamide hydrochloride 97%
(2S,5R)-5-(4-((2-Fluorobenzyl)oxy)phenyl)pyrrolidine-2-carboxamide hydrochloride

Precaución: for Research Use Only

Formula: C₁₈H₂₀ClFN₂O₂
MW: 350.82 g/mol
MDL Number: MFCD28900694
CAS Number: 934240-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR