5-(Benzyloxy)-5-oxopentanoic acid 97%

Supplier: Ambeed
Warning

A139671-100G A139671-10G A139671-1G A139671-250MG A139671-5G A139671-25G
77150-004EA 392 USD
77150-004 77150-006 77150-008 77150-010 77150-012 77900-728
5-(Benzyloxy)-5-oxopentanoic acid 97%
5-(Benzyloxy)-5-oxopentanoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₄O₄
MDL Number: MFCD00447718
CAS Number: 54322-10-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR