Ethyl-5-chloroimidazo[1,2-a]pyridine-2-carboxylate 95%

Supplier: Ambeed
Warning

A139613-100MG A139613-250MG A139613-5G A139613-1G
77149-998EA 19 USD
77149-998 77150-000 77150-002 77901-553
Ethyl-5-chloroimidazo[1,2-a]pyridine-2-carboxylate 95%
Ethyl 5-chloroimidazo[1,2-a]pyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₉ClN₂O₂
MDL Number: MFCD09864797
CAS Number: 67625-36-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR