Linsitinib 98%

Supplier: Ambeed
Warning

A139562-10MG A139562-1MG A139562-5MG A139562-1G A139562-2MG A139562-250MG A139562-50MG A139562-100MG
77149-992EA 68 USD
77149-992 77149-994 77149-996 77566-798 77611-266 77685-193 77685-194 77902-023
Linsitinib 98%
Linsitinib

Precaución: For research use only.

Formula: C₂₆H₂₃N₅O
MDL Number: MFCD12912153
CAS Number: 867160-71-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR