N-Cyclohexylhydroxylamine hydrochloride 95%

Supplier: Ambeed
Warning

A139092-1G A139092-250MG A139092-5G
77149-982EA 17 USD
77149-982 77149-984 77685-190
N-Cyclohexylhydroxylamine hydrochloride 95%
N-Cyclohexylhydroxylamine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₃NO·HCl
MW: 151.64 g/mol
MDL Number: MFCD00012565
CAS Number: 25100-12-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR